Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Sulfamidase/SGSH Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Sulfamidase/SGSH |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Sulfamidase/SGSH Polyclonal specifically detects Sulfamidase/SGSH in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Sulfamidase/SGSH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
heparan sulfate sulfatase, HSS, HSSN-sulphoglucosamine sulphohydrolase, MPS3A, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMD, SFMDEC 3.10.1.1, SGSH, Sulfoglucosamine sulfamidase, Sulphamidase | |
SGSH | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
NP_061292 | |
6448 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Sulfamidase/SGSH Arg23-Leu502 Accession # NP_061292 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Sulfamidase/SGSH in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title