Learn More
Description
Specifications
Specifications
| Assay Range | 31.3 to 2000 pg/mL |
| Assay Sensitivity | 3.7pg/mL |
| Product Type | ELISA Kit |
| Specificity | The assay detects both natural and recombinant mouse TNF alpha. The interference of circulating factors of the immune systemwas evaluated by spiking these proteins at physiologically relevant concentrations into mouse IL-10 positive serum. There was no crossreactivity detected. |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7124 |
| Gene Symbol | TNF |
| Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to mouse TNF-alpha, Biotin-Conjugate anti-mouse TNF-alpha polyclonal antibody, Streptavidin-HRP, Mouse TNF-alpha Standard lyophilized, 2ng/mL upon reconstitution, Calibrator Diluent, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
| Quantity | 96 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
