Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TNF RI/TNFRSF1A Antibody, R&D Systems™

Hamster Monoclonal Antibody has been used in 9 publications
$214.00 - $2150.00
Specifications
Antigen | TNF RI/TNFRSF1A |
---|---|
Clone | 55R170 |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation, Neutralization 5-20 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Monoclonal |
Description
TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
TNF RI/TNFRSF1A | |
Western Blot 1 ug/mL, Immunoprecipitation, Neutralization 5-20 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
7132 | |
Recombinant mouse TNF RI/TNFRSF1A Extracellular domain | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TNF RI/TNFRSF1A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse (rm) CD27, rmCD30, rhCD40, rmCD40, rhDR3, rhDR6, rmEDAR, rhFas, rmFas, rhEDAR, rhGITR, rmGITR, rhHVEM, rhLT beta R, rmLT beta R, rhNGF R, rhOPG, rmOPG, rhRANK, rmRANK, rhTAJ, rhTNF RI, rhTNF RII or rmTNF RII is observed. |
55R170 | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
Hamster | |
Mouse | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
TNFRSF1A | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title