Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TrkC Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1404
This item is not returnable.
View return policy
Description
TrkC Monoclonal specifically detects TrkC in Mouse samples. It is validated for Western Blot.Specifications
TrkC | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q6VNS1 | |
NTRK3 | |
Mouse myeloma cell line NS0-derived recombinant mouse TrkC Cys32-Thr429 Accession # Q6VNS1 | |
500 μg | |
Primary | |
Detects mouse TrkC in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50% cross-reactivity with recombinant human (rh) TrkC is observed and no cross-reactivity with rhTrkA, recombinant rat TrkA, or recombinant mouse TrkB is observed. | |
Mouse | |
Purified |
Western Blot | |
227403 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, NTRK3, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4916 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction