Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TrkC Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | TrkC |
---|---|
Clone | 227403 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
TrkC Monoclonal specifically detects TrkC in Mouse samples. It is validated for Western Blot.Specifications
TrkC | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q6VNS1 | |
4916 | |
Mouse myeloma cell line NS0-derived recombinant mouse TrkC Cys32-Thr429 Accession # Q6VNS1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TrkC in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50% cross-reactivity with recombinant human (rh) TrkC is observed and no cross-reactivity with rhTrkA, recombinant rat TrkA, or recombinant mouse TrkB is observed. |
227403 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, NTRK3, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
NTRK3 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title