Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Trypsin 3/PRSS3 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
| Antigen | Trypsin 3/PRSS3 |
|---|---|
| Clone | 419910 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB35651SP
![]() |
R&D Systems
MAB35651SP |
25 μg |
Each for $200.50
|
|
|||||
MAB35651
![]() |
R&D Systems
MAB35651 |
100 μg |
Each for $468.00
|
|
|||||
Description
Trypsin 3/PRSS3 Monoclonal specifically detects Trypsin 3/PRSS3 in Mouse samples. It is validated for Western Blot.Specifications
| Trypsin 3/PRSS3 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| NP_035775 | |
| 5646 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Trypsin 3/PRSS3 Phe16-Asn246 Accession # NP_035775 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Trypsin 3/PRSS3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, weak cross-reactivity with recombinant human (rh) Trypsin 3 is observed and no cross-reactivity with rhTrypsin 1 or rhTrypsin 2 is observed. |
| 419910 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsin, Mesotrypsinogen, MTG, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS3, PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin 4, Trypsin III, Trypsin IV, trypsin-3, Trypsinogen 15, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
| PRSS3 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title