Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse u-Plasminogen Activator (uPA)/Urokinase Antibody, R&D Systems™

Rat Monoclonal Antibody
$214.00 - $549.50
Specifications
Antigen | u-Plasminogen Activator/Urokinase |
---|---|
Clone | 901420 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9185SP
![]() |
R&D Systems
MAB9185SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9185100
![]() |
R&D Systems
MAB9185100 |
100 μg |
Each for $549.50
|
|
|||||
Description
u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
u-Plasminogen Activator/Urokinase | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
P06869 | |
5328 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
901420 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator | |
Mouse myeloma cell line NS0-derived recombinant mouse uPA Met1-Phe433 Accession # P06869 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse uPA in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title