Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse VE-Statin Isoform 1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | VE-Statin/EGFL7 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
VE-Statin/EGFL7 Polyclonal specifically detects VE-Statin/EGFL7 in Mouse samples. It is validated for Western Blot.Specifications
VE-Statin/EGFL7 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1 | |
EGFL7 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9QXT5 | |
51162 | |
E. coli-derived recombinant mouse VE-Statin Isoform 1 Thr21-Leu275 Accession # Q9QXT5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse VE-Statin Isoform 1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human VE-Statin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title