Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse VEGFR3/Flt-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7431
This item is not returnable.
View return policy
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
VEGFR3/Flt-4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, Flt-4, FLT4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2324 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
ELISA | |
102804 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q5SU94 | |
FLT4 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGFR3/Flt-4, Tyr25-Asp770, Accession # Q5SU94 | |
500 μg | |
Primary | |
Detects mouse VEGFR3/Flt-4 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) VEGFR1 or rmVEGFR2 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction