Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse VEGFR3/Flt-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | VEGFR3/Flt-4 |
---|---|
Clone | 102804 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
VEGFR3/Flt-4 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, Flt-4, FLT4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
FLT4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
102804 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q5SU94 | |
2324 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGFR3/Flt-4, Tyr25-Asp770, Accession # Q5SU94 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse VEGFR3/Flt-4 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) VEGFR1 or rmVEGFR2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title