Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Whole Serum Antibody, Novus Biologicals™
SDP

Catalog No. NBP244094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244094 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244094 Supplier Novus Biologicals Supplier No. NBP244094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Mouse Whole Serum in Mouse samples, and it is validated for ELISA, Immunoprecipitation, Western Blotting

Mouse Whole Serum Polyclonal specifically detects Mouse Whole Serum in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Mouse Whole Serum
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Gene Accession No. P07724
Gene Alias ALB, DKFZp779N1935, PRO0883, PRO0903, PRO1341, PRO2044, PRO2619, Serum albumin
Host Species Rabbit
Immunogen Mouse serum proteins (Uniprot: P07724)
Molecular Weight of Antigen 68.69 kDa
Purification Method Delipidation and Defibrination
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute with 2.0 ml deionized water (or equivalent)
Target Species Mouse
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.