Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-10b Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB2110
This item is not returnable.
View return policy
Description
Wnt-10b Monoclonal specifically detects Wnt-10b in Mouse samples. It is validated for Western Blot.Specifications
| Wnt-10b | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P48614 | |
| WNT10B | |
| E. coli-derived recombinant mouse Wnt-10b Arg263-Arg332 Accession # P48614 | |
| 100 μg | |
| Primary | |
| Detects mouse Wnt-10b in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Wnt-1, -3a, -4, -5a, -5b, -8a, -11, or -16 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 254206 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, Wnt10b, Wnt-12, WNT12 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7480 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction