Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-10b Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $502.50
Specifications
| Antigen | Wnt-10b |
|---|---|
| Clone | 254206 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
Wnt-10b Monoclonal specifically detects Wnt-10b in Mouse samples. It is validated for Western Blot.Specifications
| Wnt-10b | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P48614 | |
| 7480 | |
| E. coli-derived recombinant mouse Wnt-10b Arg263-Arg332 Accession # P48614 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Wnt-10b in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Wnt-1, -3a, -4, -5a, -5b, -8a, -11, or -16 is observed. |
| 254206 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, Wnt10b, Wnt-12, WNT12 | |
| WNT10B | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title