Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-8a Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2248SP
This item is not returnable.
View return policy
Description
Wnt-8a Monoclonal specifically detects Wnt-8a in Mouse samples. It is validated for Western Blot.Specifications
Wnt-8a | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, Wnt8a, WNT8DWNT8d | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7478 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
254126 | |
Western Blot 1 ug/mL | |
Q64527 | |
WNT8A | |
E. coli-derived recombinant mouse Wnt-8a Ala20-Trp354 (predicted) Accession # Q64527 | |
25 μg | |
Primary | |
Detects mouse Wnt-8a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Wnt-1, -3a, -4, -5a, or -10b is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction