Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-8a Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | Wnt-8a |
---|---|
Clone | 254126 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Wnt-8a Monoclonal specifically detects Wnt-8a in Mouse samples. It is validated for Western Blot.Specifications
Wnt-8a | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, Wnt8a, WNT8DWNT8d | |
WNT8A | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
254126 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q64527 | |
7478 | |
E. coli-derived recombinant mouse Wnt-8a Ala20-Trp354 (predicted) Accession # Q64527 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Wnt-8a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Wnt-1, -3a, -4, -5a, or -10b is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title