Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ZNRF3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $544.12
Specifications
| Antigen | ZNRF3 |
|---|---|
| Clone | 2035A |
| Dilution | Immunocytochemistry 5-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8328SP
![]() |
R&D Systems
MAB8328SP |
25 μg |
Each for $214.00
|
|
|||||
MAB8328100
![]() |
R&D Systems
MAB8328100 |
100 μg |
Each for $544.12
|
|
|||||
Description
ZNRF3 Monoclonal specifically detects ZNRF3 in Mouse samples. It is validated for Immunocytochemistry.Specifications
| ZNRF3 | |
| Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| Q5SSZ7 | |
| 84133 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2035A | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BK747E2.3, KIAA1133, novel C3HC4 type Zinc finger (ring finger), RNF203, zinc and ring finger 3, zinc/RING finger protein 3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ZNRF3 Lys53-Met216 Accession # Q5SSZ7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse ZNRF3 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title