Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moxonidine 98.0+%, TCI America™
SDP

Catalog No. M26601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26601G 1 g
M2660200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26601G Supplier TCI America Supplier No. M26601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75438-57-2
Molecular Formula C9H12ClN5O
Molecular Weight (g/mol) 241.68
MDL Number MFCD22689455
InChI Key WPNJAUFVNXKLIM-UHFFFAOYSA-N
Synonym 4-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine
PubChem CID 4810
ChEBI CHEBI:7009
IUPAC Name 4-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methylpyrimidin-5-amine
SMILES COC1=NC(C)=NC(Cl)=C1NC1=NCCN1

Specifications

Melting Point 206°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 241.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Moxonidine
RTECSNumber UV6260290
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.