Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2XYT-Broth, Powder

Catalog No. p-4474883
Click to view available options
Quantity:
454 g
1000 g
5000 g

Optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages

  • Contents/L: 16g tryptone, 10g yeast extract, 5g NaCl
  • Use 31g/L

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Microbiology Media
Formulation Tryptone 16g/L, Yeast extract 10g/L, NaCl 5g/L
For Use With (Application) For growth and maintenance of M13 phage or other filamentous phage
Quantity 454 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.