Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Alcohol Oxidase

Catalog No. p-3563323
Click to view available options
Quantity:
1 kU
5 kU

From Pichea pastoris E.C.1.1.3.13

Catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide. Activity: 10-20U per mg protein. Unit Definition: One unit will oxidize 1.0μmole of methanol to formaldehyde per minute at pH 7.5 at 25°C. Solution in sucrose.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥10 U/mg
Color Reddish Brown
pH 7.2
For Use With (Application) Colorimetric determination of Blood Alcohol
Storage Buffer 30% Sucrose, 0.1M Potassium Phosphate Buffer at pH 8
Content And Storage -20°C
Source Pichea pastoris
Quantity 1 kU
Product Type Alcohol Oxidase
Molecular Weight (g/mol) ≈630000
Form Frozen
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.