Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ alpha-Chymotrypsin from Bovine Pancreas

Catalog No. p-4605312
Click to view available options
Quantity:
250 mg
1 g
5 g

A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains on carboxyl end of the bond

α-Chymotrypsin preferentially catalyzes the hydrolysis of peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan.

  • CAS Number: 9004-07-3
  • EC Number: 232-671-2
  • MDL Number: MFCD00130481
  • Extinction Coefficient: 20.4
  • Format: white powder
  • From Bovine Pancreas
  • Crystallized, dialyzed, salt-free lyophilized powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1X
Color White
For Use With (Application) Catalyzes the hydrolysis of Peptide bonds involving L-isomers of Tyrosine, Phenylalanine, Tryptophan
Content And Storage 0°C
Source Bovine pancreas
Quantity 250 mg
Product Type alpha-Chymotrypsin
Molecular Weight (g/mol) 25
Inhibitors Heavy Metals, natural Trypsin inhibitors to various degrees, Organophosphorus compounds, AEBSF, alpha-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and a-2-Macroglobulin
Form Crystallized, Lyophilized
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.