Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Ascorbate Oxidase
Click to view available options
Quantity:
1 kU
200 U
400 U
Description
Activity: 200U/mg solid. Unit Definition: One unit will oxidize 1.0μmole of L-ascorbate to dehydroascorbate per minute at pH 5.6 and20 °C. Lyophilized powder.

Specifications
Specifications
Concentration | ≥200 U/mg |
Color | Light Blue |
pH | 7 to 10 |
For Use With (Application) | Enzymatic determination of Ascorbic Acid |
Content And Storage | 0°C |
Source | Cucumis species |
Quantity | 400 U |
Product Type | Ascorbate Oxidase |
Molecular Weight (g/mol) | 1320001, 140000 |
Inhibitors | Azide, Cyanide, Na2S, Diethyldithiocarbamate (Na) |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction