Learn More
MP Biomedicals™ Beta-Galactosidase from Escherichia coli Greater than 300U/mg
Shop All MP Biomedicals, Inc ProductsDescription
β-Galactosidase cleaves terminal galactose residues, which are β -1-->14 linked to a monosaccharide, glycopeptide, or oligosaccharide. The enzyme requires Mg2+ for optimal activity.
- Substrate Specificity: Enzyme specifically hydrolyzes β-D-galactosyl linkage
- Presentation: White powder
- Isoelectric point: 4.61 (Lit.)
- Optimum pH: 7.0 to 7.5, pH
- Stability: pH 6.5 to 8.5 (25°C, 20 hr)(Lit.); Thermal Stability: Below 50°C (pH 7.3, 15 min) (Lit.)
- Optimum Temperature: 50 to 55°C (Lit.)
- Unit Definition: One unit catalyzes the hydrolysis of 1 μmole of o-nitrophenyl-β-D-galactopyranoside per minute at 25°C, pH 7.5
Specifications
Specifications
| Concentration | ≥300 U/mL |
| Color | White |
| Components | Reaction mixture- 2.5mL 0.1M Phosphate Buffer (pH 7.3), 0.1mL Mercaptoethanol solution, 0.1mL MgCl2 solution , 0.2mL ONPG solution |
| pH | 7 to 7.5 |
| For Use With (Application) | Structural investigation of Carbohydrate, the determination of Lactose (food stuff analysis) and as an Enzyme label for Enzyme immunoassay |
| Content And Storage | 0°C |
| Source | E. coli |
| Quantity | 1 kU |
| Product Type | beta-Galactosidase |
| Molecular Weight (g/mol) | 540000 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.