Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Bilirubin Oxidase
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 U
100 U
Description
Activity: ≥0.8U/mg solid. Unit Definition: One unit is the amount of enzyme that oxidizes one micromole of bilirubin per minute at 37°C, pH 8.4. Lyophilized powder.

Specifications
Specifications
Concentration | ≥0.8 U/mg |
Color | Light Blue/Green |
For Use With (Application) | Degrade Bilirubin, used to determine free hemoglobin in icteric specimens, also has potential application in dye effluent decolorization and is a potential treatment for neonatal jaundice |
Content And Storage | -20°C |
Source | Myrothecium species |
Quantity | 100 U |
Product Type | Bilirubin Oxidase |
Molecular Weight (g/mol) | 52000 |
Form | Lyophilized Powder |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction