Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Catalase from Beef Liver

Catalog No. p-4605329 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g

Oxidoreductase enzyme for catalyzing conversion of hydrogen peroxide to water and oxygen, 2H2O2 → 2H2O + O2

Hydrogen peroxide is a harmful byproduct of many normal metabolic processes. to prevent damage to cells and tissues, it must be quickly converted into other, less dangerous substances. To this end, catalase is frequently used by cells to rapidly catalyze the decomposition of hydrogen peroxide into less-reactive gaseous oxygen and water molecules.

  • CAS Number: 9001-05-2
  • EC Number: 232-577-1
  • MDL Number: MFCD00081483
  • Format: Dark green powder
  • Specific Activity: ∽13,000U/mg solid
  • Extinction Coefficient: 36.5 (276nm)
  • Isoelectric point (pI): 5.4
  • Dissolves readily at 5mg/mL in 0.05M sodium/potassium phosphate (pH 7.0) giving clear green solution

Used in the food industry for removing hydrogen peroxide from milk prior to cheese production.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈13,000 U/mg
Color Dark Green
For Use With (Application) Used in the food industry for removing Hydrogen Peroxide from milk prior to cheese production
Content And Storage 0°C
Source Beef liver
Quantity 1 g
Product Type Catalase
Molecular Weight (g/mol) 250
Form Lyophilized Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.