Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Cholesterol Esterase from Pseudomonas Species
Click to view available options
Quantity:
1 kU
200 U
50 U
Description
- CAS Number: 9026-00-0
- EC Number: 232-808-6
- MDL Number: MFCD00071052
- Format: yellowish tan lyophilized powder
- Specific activity: ≥100U/mg (solid)
- Isoelectric point: 5.9 ±0.1
- Stable at -20°C for at least one year
Cholesterol Esterase is useful for enzymatic determination of total cholesterol when coupled with cholesterol oxidase in clinical analysis.

Specifications
Specifications
Concentration | ≥100 U/mg |
Color | Yellowish Tan |
pH | 7 to 9 |
For Use With (Application) | Enzymatic determination of Total Cholesterol when coupled with Cholesterol Oxidase in clinical analysis |
Content And Storage | 0°C |
Source | Pseudomonas species |
Quantity | 1 kU |
Product Type | Cholesterol Esterase |
Molecular Weight (g/mol) | ≈300000 |
Inhibitors | Hg- - Ag ionic detergents |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction