Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Cholesterol Esterase from Pseudomonas Species
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 kU
200 U
50 U
Description
- CAS Number: 9026-00-0
- EC Number: 232-808-6
- MDL Number: MFCD00071052
- Format: yellowish tan lyophilized powder
- Specific activity: ≥100U/mg (solid)
- Isoelectric point: 5.9 ±0.1
- Stable at -20°C for at least one year
Cholesterol Esterase is useful for enzymatic determination of total cholesterol when coupled with cholesterol oxidase in clinical analysis.
Specifications
Specifications
| Concentration | ≥100 U/mg |
| Color | Yellowish Tan |
| pH | 7 to 9 |
| For Use With (Application) | Enzymatic determination of Total Cholesterol when coupled with Cholesterol Oxidase in clinical analysis |
| Content And Storage | 0°C |
| Source | Pseudomonas species |
| Quantity | 50 U |
| Product Type | Cholesterol Esterase |
| Molecular Weight (g/mol) | ≈300000 |
| Inhibitors | Hg- - Ag ionic detergents |
| Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction