Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ CSM-TRP, Powdered Media
Click to view available options
Quantity:
10 g
100 g
Description
Formulation
- Add 0.74g of CSM-Trp to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium
Applications:
Yeast cell culture, Gene expression, Transcription regulation

Specifications
Specifications
Certifications/Compliance | Molecular biology Certified™ |
Content And Storage | 15°C to 30°C |
Form | Powder |
Grade | Molecular biology |
Product Type | Yeast Media |
Formulation | Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L |
For Use With (Application) | For growth of Saccharomyces cerevisiae |
Quantity | 10 g |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction