Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ CSM-TRP-URA, Powdered Media
Click to view available options
Quantity:
10 g
100 g
Description
Formulation
- Add 0.72g of CSM-Trp-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium
Applications:
Yeast cell culture, Gene expression, Transcription regulation

Specifications
Specifications
Certifications/Compliance | Molecular biology Certified™ |
Content And Storage | 15°C to 30°C |
Form | Powder |
Product Type | Yeast Media |
Formulation | Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L |
For Use With (Application) | For growth of Saccharomyces cerevisiae |
Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction