Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Deoxyribonuclease I from Bovine Pancreas

Catalog No. p-4605343 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 mg
20 mg

Used as genetic marker for forensic identification

An endonuclease that splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide. The average chain of limit digest is a tetranucleotide. DNase I acts upon single chain DNA, and upon double-stranded DNA and chromatin.

  • CAS Number: 9003-98-9
  • EC Number: 232-667-0
  • MDL Number: MFCD00130918
  • Format: White powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥2000 kU/mg
Color White
pH 7.8
For Use With (Application) Used as a genetic marker for forensic identification
Content And Storage 4°C
Source Bovine pancreas
Quantity 5 mg
Product Type Deoxyribonuclease I
Molecular Weight (g/mol) 31000
Inhibitors EDTA 7,8 and Sodium Dodecyl Sulfate
Form Lyophilized Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.