Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Diastase
Click to view available options
Quantity:
25 g
100 g
Description
Diastase, [9000-92-4]
- Activity: 1000 Lintner u/g
- Melting Point: 175°C (dec)

Specifications
Specifications
Concentration | 1000 U/g |
Color | Off-White |
For Use With (Application) | Digests 50g of Starch in <30 min. |
Content And Storage | 4°C |
Source | Malt |
Quantity | 100 g |
Product Type | Diastase |
Form | Powder |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction