Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ DOBA Medium, Powdered Media
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
454 g
1000 g
Description
Formulation
- Use 43.7g powder liter
- Contents/liter: DOB, 17g Agar-Y
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | Molecular biology |
| Product Type | Yeast Media |
| Formulation | Nitrogen Sources : Ammonium Sulfate 5g/L, Carbon Source: Dextrose 20g/L, Vitamins : Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg/L, Trace Elements: Boric Acid 0.5mg/L, Copper Sulfate 0.04mg/L, Potassium Iodide 0.1mg/L, Ferric Chloride 0.2mg/L, Manganese Sulfate 0.4mg/L, Sodium Molybdate 0.2mg/L, Zinc Sulfate 0.4mg/L, Salts: Potassium Phosphate, Monobasic 1000mg/L, Magnesium Sulfate 500mg/L, Sodium Chloride 100mg/L, Calcium Chloride 100mg/L, Agar 17g/L |
| For Use With (Application) | For Yeast cell culture, gene expression, transcription regulation |
| Quantity | 454 g |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction