Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Ficin
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Activity: 300-500 milk clotting units/mg. Hygroscopic.

Specifications
Specifications
Color | White |
pH | 5 to 7.5 |
For Use With (Application) | Cleavage of Mouse IgG |
Content And Storage | 4°C |
Source | Fig tree Ficus glabrata |
Quantity | 25 g |
Product Type | Ficin |
Molecular Weight (g/mol) | ≈25 |
Inhibitors | Iodoacetamide, Iodoacetic Acid, N-ethylmaleimide, Mercuric Chloride, DFP (Diisopropyl Fluorophosphate), TLCK (Na-p-Tosyl-lysine Chloromethyl Ketone), TPCK (N-Tosyl-L-phenylalanine Chloromethyl Ketone) |
Form | Powder |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction