Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ficin

Catalog No. p-3563472 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 g
25 g
100 g

From fig tree

Activity: 300-500 milk clotting units/mg. Hygroscopic.

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 5 to 7.5
For Use With (Application) Cleavage of Mouse IgG
Content And Storage 4°C
Source Fig tree Ficus glabrata
Quantity 5 g
Product Type Ficin
Molecular Weight (g/mol) ≈25
Inhibitors Iodoacetamide, Iodoacetic Acid, N-ethylmaleimide, Mercuric Chloride, DFP (Diisopropyl Fluorophosphate), TLCK (Na-p-Tosyl-lysine Chloromethyl Ketone), TPCK (N-Tosyl-L-phenylalanine Chloromethyl Ketone)
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.