Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ β- Galactosidase from Escherichia coli
Click to view available options
Quantity:
1 kU
10 kU
5 kU
Description
- CAS Number: 9031-11-2
- EC Number: 232-864-1
- Format: Pale gray fuffy powder
- Specific activity: ≥50U/mg
- Isoelectric point: 4.61
- Optimum pH: 7 to 7.5
Used for structural investigation of carbohydrate, determination of lactose (foodstuff analysis) and as an enzyme label for enzyme immunoassay.

Specifications
Specifications
Concentration | ≥50 U/mg |
Color | Pale Gray |
pH | 7 to 7.5 |
For Use With (Application) | Structural investigation of Carbohydrate, the determination of Lactose (food stuff analysis) and as an Enzyme label for Enzyme immunoassay |
Content And Storage | 4°C |
Source | E. coli |
Quantity | 5 kU |
Product Type | beta-Galactosidase |
Molecular Weight (g/mol) | 540000 |
Inhibitors | p-Chloromercuribenzoate, lodoacetamide, Heavy Metal Ions (Zn++, Fe++, Zn++, Cd++, Cu++, Pb++, Ag+, Hg++), Ionic Detergents SDS, DAC |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction