Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Glucose-6-Phosphate Dehydrogenase, From Yeast
Click to view available options
Quantity:
1 kU
500 U
Description
Glucose-6-Phosphate Dehydrogenase, [9001-40-5]
- G-6-PDase
- Salt-free
- Lyophilized powder
- Activity: -200 units/mg protein.
- Unit Definition: One unit will cause the reduction of 1.0 ++mole of NADP+ per minute at pH 8.0 and 25°C.

Specifications
Specifications
Concentration | ≈200 U/mg |
Color | White |
pH | 5.5 to 7.5 |
For Use With (Application) | Converts Glucose-6-phosphate into 6-phosphoglucono-d-lactone |
Content And Storage | -20°C |
Source | Yeast |
Quantity | 500 U |
Product Type | Glucose-6-phosphate Dehydrogenase |
Molecular Weight (g/mol) | 104000 |
Inhibitors | Acyl-CoA, Metal ions |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction