Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Lactoperoxidase, 60-80U/mg
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
Description
Activity: 60-80U/mg protein. A412/A280 = 0.70 to 0.85. Unit Definition: One unit will form 1.0mg of purpurogallin from pyrogallol in 20 seconds at pH 6.0 and 20°C.

Specifications
Specifications
Concentration | 60 to 80 U/mg |
Color | Brown |
For Use With (Application) | Typically used for labelling Proteins with Radioiodine |
Content And Storage | 0°C |
Source | Bovine Milk |
Quantity | 1 mg |
Product Type | Lactoperoxidase |
Molecular Weight (g/mol) | 77500 |
Inhibitors | Hydrazines, Cyanide |
Form | Lyophilized Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction