Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Linbro™ PCR Rainbow Microcentrifuge Tubes
Click to view available options
Color:
Assorted
Natural
Specifications
Specifications
Capacity (Metric) | 0.25 mL |
Color | Natural |
Material | Polypropylene |
For Use With (Application) | PCR applications |
Includes | Frosted writing area |
Packaging | 500 Case |
Bottom Shape | Conical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction