Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Liver S9 Homogenate, Suspended in Potassium Chloride

Catalog No. p-96448 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
2 mL
5 mL

Post Mitochondrial Supernatant

  • Rat Liver S9
  • Aroclor 1254 Induced
  • Suspended in potassium chloride

TRUSTED_SUSTAINABILITY

Specifications

Components 0.15M KCl
For Use With (Application) Ames mutagen assay, study of xenobiotic metabolism and drug interactions
Content And Storage -70°C
Source Male Rat, Sprague Dawley strain
Quantity 5 mL
Product Type Liver S9 Homogenate
Form Frozen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.