Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysozyme from Chicken Egg White

Catalog No. p-4397705 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g
10 g

2X crystallized, salt-free and soluble

Activity: >9000 units/mg protein. Unit Definition: One unit will cause a decrease in A450 of 0.001 per minute at pH 6.24 and 25°C using Micrococcus lysodeikticus as substrate.

  • Lyophilized
  • Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
  • CAS: 9001-63-2

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Color White
pH 11
For Use With (Application) Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen
Content And Storage 0°C
Source Chicken egg white
Quantity 5 g
Product Type Lysozyme
Molecular Weight (g/mol) 14388
Inhibitors Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes
Form Crystallized, Lyophilized
Show More Show Less

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.