Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Malate Dehydrogenase, From Porcine Heart (Mitochondrial)
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 kU
Description
Malate Dehydrogenase, [9001-64-3]
- CAS Number: 9001-64-3
- EC Number: 232-622-5
- Specific Activity: ∽1200u/mg protein
- Format: Powder
Specifications
Specifications
| Concentration | ≈1200 U/mg |
| Color | White |
| For Use With (Application) | Enzymatic determination of L-malate and Gluamate Oxaloacetate Transaminase |
| Content And Storage | 0°C |
| Source | Porcine heart |
| Quantity | 10 kU |
| Product Type | Malate Dehydrogenase |
| Form | Lyophilized Powder |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction