Learn More
MP Biomedicals™ Papain from Papaya Latex
Shop All MP Biomedicals, Inc ProductsDescription
Papain cleaves peptide bonds of basic amino acids, leucine, or glycine. It is also hydrolyzes esters and amides.
- CAS Number: 9001-73-4
- EC Number: 232-627-2
- MDL Number: MFCD00131791
- Format: Milky white solution
- Specific Activity: ≤10 units/mg Protein
- Extinction coefficient: 25.0
- Isoelectric point (pI): pH 9.6
- Optimum pH: 6.0 to 7.0
Used to produce Fab fragments of antibodies, cell dissociation, morphological isolation of intact cortical neurons from postnatal rats. Limited papaindigestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify red cell surface to enhance or destroy reactivity of many red cell antigens. Useful for platelet serology studies and has also been used in enzymatic synthesis of amino acids, peptides, and other molecules.
Specifications
Specifications
| Concentration | 2X |
| Color | Milky White |
| pH | 6 to 7 |
| For Use With (Application) | Produce Fab fragments of Antibodies, cell dissociation, morphological isolation of intact cortical neurons from Postnatal Rats |
| Content And Storage | 4°C |
| Source | Papaya Latex |
| Quantity | 1 g |
| Product Type | Papain |
| Molecular Weight (g/mol) | 23000 |
| Inhibitors | Sulfhydryl groups including heavy Metals, Carbonyl reagents, Aldehydes, Benzoylamidoacetonitrile, AEBSF, antipain, cystatin, E-64, leupeptin, PMSF, TLCK and TPCK |
| Show More |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.