Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Pectinase from Aspergillus niger
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 kU
10 kU
100 kU
25 kU
5 kU
Description
Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.
- Lyophilized
- Activity: >1000 u/gm

Specifications
Specifications
Concentration | >1000 U/g |
Color | Beige |
For Use With (Application) | Used in plant protoplast preparation to digest cell wall prior to organelle isolation |
Content And Storage | 4°C |
Source | Aspergillus niger |
Quantity | 10 kU |
Product Type | Pectinase |
Form | Lyophilized Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction