Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Peroxidase, 250-330U/mg solid

Catalog No. p-3563592 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 kU
10 kU
25 kU
5 kU

From horseradish

Activity: 250-330U/mg solid. Unit Definition: One unit will form 1mg purpurogallin in 20 seconds from pyrogallol at pH 6.0 and 20°C. Essentially salt-free powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥250 U/mg
Color Reddish Brown
pH 7
For Use With (Application) ELISA, immunohistochemistry, western blotting
Content And Storage 0°C
Source Horseradish
Quantity 5 kU
Product Type Peroxidase
Molecular Weight (g/mol) ≈44000
Inhibitors HRP is reversibly inhibited by Cyanide and Sulfide at a concentration of 10-5M, p-Aminobenzoic Acid, Azide, Cyclopropanone, L-cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite and Vanadate and divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb
Form Crystalline Powder
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.