Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoglucose Isomerase from Baker's Yeast, MP Biomedicals™

Catalog No. ICN10098305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10098305 5 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10098305 Supplier MP Biomedicals, Inc Supplier No. 0210098305
Add to Cart
Add to Cart

≥300 u/mg protein, Milky Suspension

  • Phosphoglucose Isomerase is used in sugar assays to convert fructose to glucose.
  • There is evidence that phosphoglucose isomerase acts as a molecular messenger.
  • It is produced and secreted by white blood cells, and acts to regulate the growth of several different cell types.
  • Phosphoglucose Isomerase(PGI) is an enzyme crucial for the interconversion of D-glucose 6 phosphate and D-fructose 6-phosphate.
  • PGI is responsible for the second step of glycolysis and is involved in glucogenesis.
  • It is highly conserved in bacteria and eukaryotes.
  • Phosphoglucose Isomerase fuctions as an isomerase, neuroleukin, autocrine motility factor, and a differentiation and maturation mediator.
TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥300 U/mg
Color Milky
pH 7 to 7.6
For Use With (Application) Used in sugar assays to convert Fructose to Glucose, acts as a molecular messenger and regulates the growth of several different cell types
Content And Storage 4°C
Source Baker's yeast
Quantity 5 kU
Product Type Phosphoglucose Isomerase
Molecular Weight (g/mol) 145000
Inhibitors Pyridoxal-5'-Phosphate
Synonym E.C.5.3.1.9
Form Suspension
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.