Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pyruvate Kinase from Rabbit Muscle

Catalog No. p-3573436 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100,000 U
25,000 U
50,000 U
5000 U

≥200u/mg protein, White Crystalline Powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥200 U/mg
Color Clear
pH 7.5
For Use With (Application) Used as a coupling Enzyme in conjunction with Lactic Dehydrogenase in quantifying ADP and the activity of Enzymes that catalyze the formation of ADP
Content And Storage -20°C
Source Rabbit muscle
Quantity 5000 U
Product Type Pyruvate Kinase
Molecular Weight (g/mol) 237000
Inhibitors Activity is inhibited by Ca2+, Fluorophosphate inhibits the Enzyme reaction competitively with respect to PEP , ATP inhibits the reaction by removal of Mg2+ from the Substrate MgADP-
Form Lyophilized Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.