Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Superoxide Dismutase

Catalog No. p-3563642 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
15 kU
3 kU
30 kU
75 kU

Catalog No. ICN19011703

Supplier: MP Biomedicals™ 0219011703

Only null left
Add to Cart
Add to Cart

From bovine erthrocytes

Activity: 3500U/mg. Unit Definition: That amount of enzyme causing a 50% inhibition in the rate of assay based on the method of McCord, J.M. and Fridovich, J. Biol. Chem., 1: 6049, (1969). Lyophilized salt-free powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥0.900 mg protein/mg solid
Color Pale Blue
For Use With (Application) Superoxide Dismutase plays a critical role in the defense of cells against the toxic effects of Oxygen radicals
Content And Storage -20°C
Source Bovine erythrocytes
Quantity 3 kU
Product Type Superoxide Dismutase
Molecular Weight (g/mol) ≈32500
Form Lyophilized Powder
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.