Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Trypsin from Beef Pancreas
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg
10 g
Concentration:
2.5% w/v
2X
Source:
Beef Pancreas
Human/Porcine/Bovine pancreas
Specifications
Specifications
| Concentration | 2.5% w/v |
| Color | White |
| For Use With (Application) | For the removal of adherent cells from a culture surface |
| Source | Beef Pancreas |
| Quantity | 10 g |
| Product Type | Trypsin |
| Molecular Weight (g/mol) | 23000 or 22000 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction