Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MPC 6827 hydrochloride
Inhibitor of microtubule polymerization; antimitotic and antitumor
Specifications
Purity | >98% |
---|---|
Product Type | MPC 6827 hydrochloride |
Molecular Weight (g/mol) | 315.8 |
Inhibitors | MPC 6827 hydrochloride |
Description
Potent inhibitor of microtubule formation (IC50 = 1.5 - 3.4 nM); inhibits polymerization of tubulin in vitro and disrupts microtubule formation in several cancer cell lines. Inhibits tumor growth in vitro and in vivo; exhibits pro-apoptotic characteristics. Brain penetrant.Specifications
>98% | |
MPC 6827 hydrochloride | |
315.8 | |
MPC 6827 hydrochloride |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title