Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPDU1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB044131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044131 Supplier Novus Biologicals Supplier No. NBP297487B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MPDU1 Polyclonal specifically detects MPDU1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MPDU1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CDGIF, FLJ14836, HBeAg-binding protein 2 binding protein A, Lec35, mannose-P-dolichol utilization defect 1, mannose-P-dolichol utilization defect 1 protein, My008, PP3958, PQLC5, SL15HBEBP2BPA, Suppressor of Lec15 and Lec35 glycosylation mutation homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MPDU1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9526
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.