Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mPEG, Mouse anti-All, Clone: 15-2b, Millipore Sigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS2002100UG
Description
Anti-mPEG, clone 15-2b, Cat. No. MABS2002, is a mouse monoclonal antibody that detects methoxy-PEG (mPEG)-conjugated proteins and has been tested for use in ELISA.
Polyethylene glycols (PEG) are non-toxic, uncharged, hydrophilic long chain polymers with a big exclusion volume in aqueous solutions. The covalent attachment of PEG is commonly employed to modify proteins and drugs that can increase their bioavailability and reduce immunogenicity and proteolytic degradation. The monomethoxylated form of polyethylene glycol (mPEG) has been widely employed in drug modifications. mPEG modification reduces nonspecific binding and prevents recognition by the reticuloendothelial system. This monoclonal antibody (clone 15-2b) specifically binds to the terminal methoxy group, but not the backbone of PEG. It displays high affinity for the terminal methoxy group and can be easily used as a detection antibody for the quantification of mPEG by ELISA. It is also suitable for detection of PEGylated liposomes and nanoparticles. The mPEG cell-based competition ELISA can detect lower molecular-weight (
Specifications
| mPEG | |
| Monoclonal | |
| Unconjugated | |
| Methoxy polyethylene glycol | |
| Methoxy PEG covalently conjugated to E.coli beta-glucuronidase. | |
| 100 μg | |
| Signaling | |
| Clone 15-2b specifically detects proteins conjugated with methoxy polyethylene glycol (mPEG). | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| IgG2b κ |
| ELISA | |
| 15-2b | |
| Purified mouse monoclonal antibody IgG2b in PBS with 0.04% sodium azide. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction